2'-Deoxyinosin ≥98%

Lieferant: Thermo Scientific

Synonyme: 9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

H52292.MD H52292.03 H52292.06
H52292.MDEA 21.9 EUR
H52292.MD H52292.03 H52292.06
2'-Deoxyinosin ≥98%
2'-Deoxyinosin

Purity: 98+% CAS No.: 890-38-0 Molecular Formula: C10H12N4O4 Formula Weight: 252.23 MDL No.: MFCD00005762

Formel: C₁₀H₁₂N₄O₄
Molecular Weight: 252,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005762
CAS-Nummer: 890-38-0
EINECS: 212-964-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR