Puromycin Dihydrochlorid

Lieferant: Invivogen
Achtung

ANT-PR-1 ANT-PR-5
INVVANT-PR-1EA 260 EUR
INVVANT-PR-1 INVVANT-PR-5
Puromycin Dihydrochlorid
Puromycin Dihydrochlorid
Formel: C₂₂H₂₉N₇O₅·2HCl
Lagertemperatur: Gefrierschrank
MDL: MFCD00012691
CAS-Nummer: 58-58-2
EINECS: 200-387-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR