2'-Desoxyadenosin Monohydrat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate
A11166.03
A11166.06
A11166.14
A11166.03EA
43.5
EUR
A11166.03
A11166.06
A11166.14
2'-Desoxyadenosin Monohydrat ≥99%
2'-Desoxyadenosin Monohydrat
Purity: 99%
CAS No.: 16373-93-6
Molecular Formula: C10H13N5O3.H2O
Formula Weight: 269.26 (251.24anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 186-189°
Beilstein Reference: 5191174
Optical Rotation: -25° (c=0.5 water)
MDL No.: MFCD00149364
Formel:
C₁₀H₁₃N₅O₃·1H₂O Molecular Weight: 269.26 g/mol (251.24 anhy) Schmelzpunkt: 186…189 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00149364 CAS-Nummer: 16373-93-6 EINECS: 213-488-7 |