2-Iodadenosin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2R,3R,4S,5R)-2-(6-Amino-2-iodo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
M-Clarity™ Qualitätsstufe = MQ100
663115-1G
663115-250MG
SIAL663115-1GEA
253
EUR
SIAL663115-1G
SIAL663115-250MG
2-Iodadenosin, Sigma-Aldrich®
2-Iodadenosin
Formel:
C₁₀H₁₂IN₅O₄ |
MDL:
MFCD01320413 CAS-Nummer: 35109-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...