Adenosin-5'-diphosphat Dinatriumsalz (ADP Dinatriumsalz)

Lieferant: Serva

Synonyme: Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate

10800.02 10800.03
SERA10800.02EA 0 EUR
SERA10800.02 SERA10800.03
Adenosin-5'-diphosphat Dinatriumsalz (ADP Dinatriumsalz)
Adenosin-5'-diphosphat Dinatriumsalz (ADP Dinatriumsalz)
Formel: C₁₀H₁₃N₅Na₂O₁₀P₂
Molecular Weight: 471,167 g/mol
MDL: MFCD00066635
CAS-Nummer: 16178-48-6
EINECS: 240-314-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR