Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Lieferant: Serva
Synonyme:
((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt
10920.01
10920.02
10920.03
10920.04
SERA10920.01EA
0
EUR
SERA10920.01
SERA10920.02
SERA10920.03
SERA10920.04
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Formel:
C₁₀H₁₄N₅Na₂O₁₃P₃ Molecular Weight: 551,14 g/mol |
MDL:
MFCD00080339 CAS-Nummer: 987-65-5 EINECS: 213-579-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...