Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate
M-Clarity™ Qualitätsstufe = MQ200
A6419-10G A6419-1G A6419-5G
SIALA6419-10GEA 380 EUR
SIALA6419-10G SIALA6419-1G SIALA6419-5G
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat, Sigma-Aldrich®
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat
Formel: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
Molecular Weight: 551.14 g/mol (anhy)
MDL: MFCD00150755
CAS-Nummer: 34369-07-8
EINECS: 213-579-1