Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate
A7699-10G A7699-1G A7699-5G
SIALA7699-10GEA 1270 EUR
SIALA7699-10G SIALA7699-1G SIALA7699-5G
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat, Sigma-Aldrich®
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat
Formel: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
Molecular Weight: 551,14 g/mol (anhy)
MDL: MFCD00150755
CAS-Nummer: 34369-07-8
EINECS: 213-579-1