Inosin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

A14459.06 A14459.14 A14459.22
A14459.06EA 24.1 EUR
A14459.06 A14459.14 A14459.22
Inosin ≥98%
Inosin

Applications: Suppresses the increase of glucose and insulin used in blood.

Formel: C₁₀H₁₂N₄O₅
Molecular Weight: 268.23 g/mol
Schmelzpunkt: 212…215 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066770
CAS-Nummer: 58-63-9
EINECS: 200-390-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR