3'-Desoxythymidin (2',3'-Didesoxythymidin), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3'-Deoxythymidine, 1-((2R,5S)-5-(Hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

M-Clarity™ Qualitätsstufe = MQ100

323616-100MG 323616-25MG
SIAL323616-100MGEA 183 EUR
SIAL323616-100MG SIAL323616-25MG
3'-Desoxythymidin (2',3'-Didesoxythymidin), Sigma-Aldrich®
3'-Desoxythymidin (2',3'-Didesoxythymidin)
Formel: C₁₀H₁₄N₂O₄
Molecular Weight: 226,23 g/mol
MDL: MFCD00010570
CAS-Nummer: 3416-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR