Thymidin (2'-Deoxythymidin), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
M-Clarity™ Qualitätsstufe = MQ200
89270-1G
89270-25G
89270-5G
SIAL89270-1GEA
77
EUR
SIAL89270-1G
SIAL89270-25G
SIAL89270-5G
Thymidin (2'-Deoxythymidin), Sigma-Aldrich®
Thymidin (2'-Deoxythymidin)
Formel:
C₁₀H₁₄N₂O₅ Molecular Weight: 242,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006537 CAS-Nummer: 50-89-5 EINECS: 200-070-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...