Thymidine 5′-triphosphate sodium salt (dTTP Na), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Thymidin-5'-triphosphorsäure Natriumsalz, 2'-Desoxythymidin-5'-triphosphorsäure Natriumsalz

T0251-10MG T0251-250MG T0251-25MG T0251-50MG
SIALT0251-10MGEA 107 EUR
SIALT0251-10MG SIALT0251-250MG SIALT0251-25MG SIALT0251-50MG
Thymidine 5′-triphosphate sodium salt (dTTP Na), Sigma-Aldrich®
Thymidine 5′-triphosphate sodium salt (dTTP Na)
Formel: C₁₀H₁₆N₂NaO₁₄P₃
Molecular Weight: 504,15 g/mol
MDL: MFCD00080338
CAS-Nummer: 18423-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR