Thymidin-5'-triphosphorsäure Tetranatriumsalz (2'-Desoxythymidin-5'-triphosphorsäure Tetranatriumsalz)

Lieferant: Serva

Synonyme: dTTP-Na4, 2'-Deoxythymidine-5'-triphosphoric acid tetrasodium salt, Thymidine 5′-triphosphate tetrasodium salt

39703.01 39703.02
SERA39703.01EA 0 EUR
SERA39703.01 SERA39703.02
Thymidin-5'-triphosphorsäure Tetranatriumsalz (2'-Desoxythymidin-5'-triphosphorsäure Tetranatriumsalz)
Thymidin-5'-triphosphorsäure Tetranatriumsalz (2'-Desoxythymidin-5'-triphosphorsäure Tetranatriumsalz)
Formel: C₁₀H₁₃N₂Na₄O₁₄P₃
Molecular Weight: 570,10 g/mol
MDL: MFCD00080338
CAS-Nummer: 3624-46-2
EINECS: 222-826-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR