2',3'-Didesoxycytidin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Zalcitabine
L10619.MF
L10619.MD
L10619.MFEA
84.4
EUR
L10619.MF
L10619.MD
2',3'-Didesoxycytidin ≥98%
2',3'-Didesoxycytidin
Purity: 98+%
CAS No.: 7481-89-2
Molecular Formula: C9H13N3O3
Formula Weight: 211.22
Storage Temperature: 2°C to 8°C
Melting Point: 216-220°
Beilstein Reference: 654956
Merck Reference: 14,10109
Optical Rotation: +90° (c=0.5 in water)
MDL No.: MFCD00012188
Formel:
C₉H₁₃N₃O₃ Molecular Weight: 211,22 g/mol Schmelzpunkt: 216…220 °C |
MDL:
MFCD00012188 CAS-Nummer: 7481-89-2 Merck Index: 14,10109 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...