2',3'-Didesoxycytidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Zalcitabine

M-Clarity™ Qualitätsstufe = MQ200

D5782-100MG D5782-250MG D5782-500MG
SIALD5782-100MGEA 377 EUR
SIALD5782-100MG SIALD5782-250MG SIALD5782-500MG
2',3'-Didesoxycytidin, Sigma-Aldrich®
2',3'-Didesoxycytidin
Formel: C₉H₁₃N₃O₃
Molecular Weight: 211,22 g/mol
MDL: MFCD00012188
CAS-Nummer: 7481-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR