2'-Desoxycytidin 99+%
Lieferant: Thermo Fisher Scientific
Synonyme:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, deoxyC
226720010
226722500
226725000
226720050
226720250
ACRO226720010EA
75.7
EUR
ACRO226720010
ACRO226722500
ACRO226725000
ACRO226720050
ACRO226720250
2'-Desoxycytidin 99+%
2'-Desoxycytidin
Formel:
C₉H₁₃N₃O₄ Molecular Weight: 227.22 g/mol Schmelzpunkt: 209…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006547 CAS-Nummer: 951-77-9 EINECS: 213-454-1 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤8 % |
HPLC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...