Nystatin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Fungicidin, Mycostatin

M-Clarity™ Qualitätsstufe = MQ200

N1638-20ML N1638-100ML
SIALN1638-20MLEA 28.7 EUR
SIALN1638-20ML SIALN1638-100ML
Nystatin, Sigma-Aldrich®
Nystatin
Formel: C₄₇H₇₅NO₁₇
Molecular Weight: 926.09 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD25541651
CAS-Nummer: 1400-61-9
EINECS: 215-749-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR