Nystatin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Fungicidin, Mycostatin
M-Clarity™ Qualitätsstufe = MQ200
N3503-5MU
N3503-25MU
SIALN3503-5MUEA
113
EUR
SIALN3503-5MU
SIALN3503-25MU
Nystatin, Sigma-Aldrich®
Nystatin
Formel:
C₄₇H₇₅NO₁₇ Molecular Weight: 926,09 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD25541651 CAS-Nummer: 1400-61-9 EINECS: 215-749-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...