Methotrexat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: L-Amethopterin hydrate, Methylaminopterin hydrate, MTX hydrate, (4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzoyl)-L-glutamic acid hydrate(1:x), Antifolan hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, (+)-Amethopterin hydrate, L(+)-Amethopterin hydrate, 4-Amino-10-methylfolic acid hydrate

M-Clarity™ Qualitätsstufe = MQ200

06563-100MG 06563-25MG 06563-5MG
SIAL06563-100MGEA 249 EUR
SIAL06563-100MG SIAL06563-25MG SIAL06563-5MG
Methotrexat Hydrat, Sigma-Aldrich®
Methotrexat Hydrat
Formel: C₂₀H₂₂N₈O₅·xH₂O
Molecular Weight: 454,44 g/mol (anhy)
MDL: MFCD00150847
CAS-Nummer: 133073-73-1
EINECS: 200-413-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR