n-Decyl-β-D-maltosid, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside
252718-1
252718-5
252718-1EA
257
EUR
252718-1
252718-5
n-Decyl-β-D-maltosid, Millipore®
n-Decyl-β-D-maltosid
Formel:
C₂₂H₄₂O₁₁ Molecular Weight: 482.56 g/mol |
MDL:
MFCD00061624 CAS-Nummer: 82494-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...