n-Decyl-β-D-maltosid

Lieferant: Uptima

Synonyme: DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside

UP426131 UP426132
UPTIUP426131EA 0 EUR
UPTIUP426131 UPTIUP426132
n-Decyl-β-D-maltosid
n-Decyl-β-D-maltosid
Formel: C₂₂H₄₂O₁₁
Molecular Weight: 482.56 g/mol
MDL: MFCD00061624
CAS-Nummer: 82494-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR