n-Dodecyl-β-D-maltosid
Lieferant: Uptima
|
Achtung
|
Synonyme:
DDM
UP346751
UP346752
UPTIUP346751EA
0
EUR
UPTIUP346751
UPTIUP346752
n-Dodecyl-β-D-maltosid
n-Dodecyl-β-D-maltosid
Formel:
C₂₄H₄₆O₁₁ Molecular Weight: 510,6 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00043012 CAS-Nummer: 69227-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...