Rhodamin B Isothiocyanat Gemisch aus Isomeren, Pulver, BioReagent geeignet zum Markieren von Eiweissen, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
RBITC
Rhodamine B isothiocyanate (RhBITC) is an organic fluorescence dye and is widely used as a fluorescent tag in a variety of biological applications.
RhBITC is adsorbed onto Ag on silica or polystyrene beads. It exhibits not only a strong surface-enhanced Raman scattering (SERS) signal but also a measurable amount of fluorescence. The RhBITC-modified Ag-deposited silica or polystyrene beads disperse well in ethanol. They are also readily coated in water with polyelectrolytes for their further derivatization with biological molecules of interest that can bind to target molecules.
Rhodamine B isothiocyanate is used for the following applications:
• Rhodamine isothiocyanate labeling
• Rhodamine-labeled synthetic APF
• Preparations of protein and fluorescent conjugates
• Fluorescent red dye
Formel:
C₂₉H₃₀ClN₃O₃S Molecular Weight: 536,08 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00136007 CAS-Nummer: 36877-69-7 EINECS: 253-248-9 UN: 3335 ADR: 9,III |
Spezifikation Testergebnisse
labeling efficiency with bovine albumin | > 70% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...