3,5-Di-tert-butyl-4-hydroxybenzaldehyd Hemihydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
140406-100G
140406-25G
SIAL140406-100GEA
122
EUR
SIAL140406-100G
SIAL140406-25G
3,5-Di-tert-butyl-4-hydroxybenzaldehyd Hemihydrat, Sigma-Aldrich®
3,5-Di-tert-butyl-4-hydroxybenzaldehyd Hemihydrat
Formel:
C₁₅H₂₂O₂·0.5H₂O |
CAS-Nummer:
207226-32-2 EINECS: 216-592-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...