3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Monohydrat
Lieferant: Cayman Chemical
Synonyme: Dihydrochloride TMB, TMB-HCl
70450-250 70450-1 70450-5
CAYM70450-250EA 73 EUR
CAYM70450-250 CAYM70450-1 CAYM70450-5
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Monohydrat
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel: C₁₆H₂₀N₂·2HCl·xH₂O
Molecular Weight: 313.27 g/mol (anhy)
MDL: MFCD00150104
CAS-Nummer: 207738-08-7
EINECS: 264-769-6