3-(2-Furyl)acrylsäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Furan-2-yl)acrylic acid
B21653.14
B21653.22
B21653.30
B21653.14EA
40.2
EUR
B21653.14
B21653.22
B21653.30
3-(2-Furyl)acrylsäure ≥99%
3-(2-Furyl)acrylsäure
Purity: 99%
CAS No.: 539-47-9
Molecular Formula: C7H6O3
Formula Weight: 138.12
Melting Point: 140-144°
Boiling Point: 286°
Merck Reference: 14,4297
MDL No.: MFCD00003257
Formel:
C₇H₆O₃ Molecular Weight: 138,12 g/mol Siedepunkt: 286 °C Schmelzpunkt: 140…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003257 CAS-Nummer: 539-47-9 EINECS: 208-718-8 Merck Index: 14,4297 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...