3-(3-Furyl)acrylsäure ≥98%

Lieferant: Thermo Scientific
Achtung

L13143.03 L13143.06 L13143.14
L13143.03EA 35.4 EUR
L13143.03 L13143.06 L13143.14
3-(3-Furyl)acrylsäure ≥98%
3-(3-Furyl)acrylsäure

Purity: 98% CAS No.: 39244-10-5 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 152-154° Beilstein Reference: 1306450

Formel: C₇H₆O₃
Molecular Weight: 138,12 g/mol
Schmelzpunkt: 152…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075074
CAS-Nummer: 39244-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR