5-Iod-1,3-dimethyluracil ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

B25173.03 B25173.06
B25173.03EA 61.2 EUR
B25173.03 B25173.06
5-Iod-1,3-dimethyluracil ≥99%
5-Iod-1,3-dimethyluracil

Purity: 99% CAS No.: 40738-83-8 Molecular Formula: C6H7IN2O2 Formula Weight: 266.04 Melting Point: 225° dec. MDL No.: MFCD00192228

Formel: C₆H₇IN₂O₂
Molecular Weight: 266.04 g/mol
Schmelzpunkt: 225 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00192228
CAS-Nummer: 40738-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR