Chlorpentamethyldisilan

Lieferant: CHEMPUR SILANE

Synonyme: 1-Chloro-1,1,2,2,2-pentamethyldisilane

SILS04440.5 SILS04440.25
SILNSILS04440.5EA 0 EUR
SILNSILS04440.5 SILNSILS04440.25
Chlorpentamethyldisilan
Chloropentamethyldisilane
Formel: C₅H₁₅ClSi₂
Molecular Weight: 166.7966 g/mol
MDL: MFCD00456692
CAS-Nummer: 1560-28-7
EINECS: 216-330-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR