1,3-Bis(2,4,6-trimethylphenyl)imidazoliumchlorid ≥97%

Lieferant: Strem Chemicals, Inc.

Synonyme: 1,3-Dimesityl-1H-imidazol-3-ium chloride, 1,3-Dimesitylimidazolium chloride

07-0299-1G 07-0299-5G
STRE07-0299-1GEA 134 EUR
STRE07-0299-1G STRE07-0299-5G
1,3-Bis(2,4,6-trimethylphenyl)imidazoliumchlorid ≥97%
1,3-Bis(2,4,6-trimethylphenyl)imidazoliumchlorid

NHC

Formel: C₂₁H₂₅ClN₂
Molecular Weight: 340,89 g/mol
MDL: MFCD02684541
CAS-Nummer: 141556-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR