4,4'-Dimethyl-2,2'-bipyridin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4,4'-Dimethyl-2,2'-bipyridine
M-Clarity™ Qualitätsstufe = MQ100
569593-1G
569593-5G
SIAL569593-1GEA
32.4
EUR
SIAL569593-1G
SIAL569593-5G
4,4'-Dimethyl-2,2'-bipyridin, Sigma-Aldrich®
4,4'-Dimethyl-2,2'-bipyridyl
Formel:
C₁₂H₁₂N₂ Molecular Weight: 184.24 g/mol |
MDL:
MFCD00006441 CAS-Nummer: 1134-35-6 EINECS: 214-483-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...