Ammoniumcer(IV)nitrat, reinst
Lieferant: Bernd Kraft
|
Gefahr
|
Synonyme:
Diammoniumhexanitratocerat, di-Ammonium-hexanitratocerat (IV), Cer(IV)ammoniumnitrat
21405.0600
21405.2600
KRAF21405.0600EA
111
EUR
KRAF21405.0600
KRAF21405.2600
Ammoniumcer(IV)nitrat, reinst
Ammoniumcer(IV)nitrat
Formel:
(NH₄)₂Ce(NO₃)₆ Molecular Weight: 548,22 g/mol Schmelzpunkt: 107…108 °C Dichte: 2,2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151121 CAS-Nummer: 16774-21-3 EINECS: 240-827-6 UN: 1477 ADR: 5.1,III Merck Index: 13,00509 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...