2-Methylbenzimidazol

Lieferant: Thermo Scientific

Synonyme: 2-Methyl-1H-benzimidazol

SB01273.1 SB01273.10 SB01273.50
MAYBSB01273.1EA 0 EUR
MAYBSB01273.1 MAYBSB01273.10 MAYBSB01273.50
2-Methylbenzimidazol
2-Methylbenzimidazol
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
Siedepunkt: 339 °C (1013 hPa)
Schmelzpunkt: 175 °C
MDL: MFCD00005598
CAS-Nummer: 615-15-6
EINECS: 210-411-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR