4-Phenoxybenzylbromid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-4-phenoxybenzene

H33895.MD H33895.03
H33895.MDEA 107 EUR
H33895.MD H33895.03
4-Phenoxybenzylbromid ≥97%
4-Phenoxybenzylbromid
Formel: C₁₃H₁₁BrO
Molecular Weight: 263,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038693
CAS-Nummer: 36881-42-2
EINECS: 253-253-6
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR