3-Aminobenzamid
Lieferant: Cayman Chemical
Synonyme:
3-Aminobenzoesäureamid
10397-1
10397-5
CAYM10397-1EA
46
EUR
CAYM10397-1
CAYM10397-5
3-Aminobenzamid
3-Aminobenzamid
Formel:
C₇H₈N₂O Molecular Weight: 136,15 g/mol Siedepunkt: 341 °C (1013 hPa) Schmelzpunkt: 111…115 °C |
MDL:
MFCD00007989 CAS-Nummer: 3544-24-9 EINECS: 222-586-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...