3-Nitro-o-phenylendiamin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Nitrobenzene-1,2-diamine, 3-Nitro-1,2-phenylenediamine
B20201.03
B20201.06
B20201.14
B20201.03EA
53.4
EUR
B20201.03
B20201.06
B20201.14
3-Nitro-o-phenylendiamin 98%
3-Nitro-o-phenylendiamin
Purity: 98%
CAS No.: 3694-52-8
Molecular Formula: C6H7N3O2
Formula Weight: 153.14
Melting Point: 157-162°
Beilstein Reference: 909344
Formel:
C₆H₇N₃O₂ Molecular Weight: 153,14 g/mol Schmelzpunkt: 157…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007722 CAS-Nummer: 3694-52-8 EINECS: 223-013-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...