(3,5-Difluorphenyl)acetonitril

Lieferant: Thermo Scientific

Synonyme: 3,5-Difluorbenzylcyanid

SB01685.1 SB01685EA
MAYBSB01685.1EA 0 EUR
MAYBSB01685.1 MAYBSB01685EA
(3,5-Difluorphenyl)acetonitril
(3,5-Difluorphenyl)acetonitril
Formel: C₈H₅F₂N
Molecular Weight: 153,13 g/mol
Siedepunkt: 221 °C (1013 hPa)
Schmelzpunkt: 34…36 °C
MDL: MFCD00061278
CAS-Nummer: 122376-76-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR