Ammoniumcer(IV)sulfat Dihydrat, reinst

Lieferant: Bernd Kraft

Synonyme: Cer(IV)ammoniumsulfat Dihydrat, tetra-Ammoniumcertetrakis(sulfat) Dihydrat

21407.0600 21407.2600
KRAF21407.0600EA 137 EUR
KRAF21407.0600 KRAF21407.2600
Ammoniumcer(IV)sulfat Dihydrat, reinst
Ammoniumcer(IV)sulfat Dihydrat
Formel: (NH₄)₂Ce(SO₄)₄·2H₂O
Molecular Weight: 632.55 g/mol
Schmelzpunkt: 130
MDL: MFCD00149417
CAS-Nummer: 10378-47-9
EINECS: 231-567-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR