Tetrahydro-4H-pyran-4-on

Lieferant: COMBI-BLOCKS

Synonyme: Tetrahydro-γ-pyran-4-on, Tetrahydro-2H-pyran-4-on, Tetrahydro-α-pyran-4-on

AM-1010-5G AM-1010-25G AM-1010-100G
COBBAM-1010-5GEA 25.5 EUR
COBBAM-1010-5G COBBAM-1010-25G COBBAM-1010-100G
Tetrahydro-4H-pyran-4-on
Tetrahydro-4H-pyran-4-on
Formel: C₅H₈O₂
Molecular Weight: 100,12 g/mol
Siedepunkt: 166 °C (1013 hPa)
Dichte: 1,08 g/cm³ (20 °C)
MDL: MFCD00006581
CAS-Nummer: 29943-42-8
EINECS: 249-967-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR