4'-Bromvalerophenon

Lieferant: Thermo Scientific

BTB12519.1 BTB12519.10 BTB12519.50
MAYBBTB12519.1EA 0 EUR
MAYBBTB12519.1 MAYBBTB12519.10 MAYBBTB12519.50
4'-Bromvalerophenon
4'-Bromvalerophenon
Formel: C₁₁H₁₃BrO
Molecular Weight: 241,13 g/mol
MDL: MFCD00000107
CAS-Nummer: 7295-44-5
EINECS: 230-729-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR