3-Amino-1H-indazol 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Amino-1H-indazol, Indazol-3-ylamin, 1H-Indazol-3-ylamin, 1H-Indazol-3-amin, Indazol-3-amin

H33435.03 H33435.09
H33435.03EA 59.6 EUR
H33435.03 H33435.09
3-Amino-1H-indazol 97%
3-Amino-1H-indazole
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00182045
CAS-Nummer: 874-05-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR