5-Bromthiazol 97%

Lieferant: Thermo Scientific
Gefahr

H55387.03 H55387.06
H55387.03EA 87 EUR
H55387.03 H55387.06
5-Bromthiazol 97%
5-Bromothiazole

Purity: 98% CAS No.: 3034-55-7 Molecular Formula: C3H2BrNS Formula Weight: 164.02 Storage Temperature: -30°C to -10°C Flash Point: 76°(169°F) Refractive Index: 1.5955 MDL No.: MFCD07787394

Formel: C₃H₂BrNS
Molecular Weight: 164,03 g/mol
Dichte: 1,835
Flammpunkt: 76 °C (169 °F)
MDL: MFCD07787394
CAS-Nummer: 3034-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR