16-Hydroxypalmitinsäure
Lieferant: Cayman Chemical
Synonyme:
16-Hydroxyhexadecansäure
9000789-1
9000789-5
CAYM9000789-1EA
63
EUR
CAYM9000789-1
CAYM9000789-5
16-Hydroxypalmitinsäure
16-Hydroxypalmitinsäure
Formel:
C₁₆H₃₂O₃ Molecular Weight: 272,43 g/mol Siedepunkt: 402 °C (1013 hPa) Schmelzpunkt: 95 °C |
MDL:
MFCD00002750 CAS-Nummer: 506-13-8 EINECS: 208-028-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...