2-Hydroxybenzimidazol
Lieferant: Thermo Scientific
Synonyme:
2-Benzimidazolinon, 2(1H)-Benzimidazolon, 2-Hydroxy-1H-benzimidazol
SB01274.1
SB01274.50
MAYBSB01274.1EA
0
EUR
MAYBSB01274.1
MAYBSB01274.50
2-Hydroxybenzimidazol
2-Hydroxybenzimidazole
Formel:
C₇H₆N₂O Schmelzpunkt: 320…322 °C Dichte: 1,42 g/cm³ (20 °C) |
MDL:
MFCD00127894 CAS-Nummer: 615-16-7 EINECS: 210-412-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...