(R)-(+)-2-Brom-α-methylbenzylalkohol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-(+)-1-(2-Bromophenyl)ethanol, (R)-2-Bromo-alpha-methylbenzyl Alcohol

H56003.03 H56003.06
H56003.03EA 51.1 EUR
H56003.03 H56003.06
(R)-(+)-2-Brom-α-methylbenzylalkohol ≥98%
(R)-(+)-2-Brom-α-methylbenzylalkohol

Purity: 98% CAS No.: 76116-20-6 Molecular Formula: C8H9BrO Formula Weight: 201.06 Melting Point: 54-56° Optical Rotation: +54° (c=1 in chloroform)

Formel: C₈H₉BrO
Molecular Weight: 201.06 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216591
CAS-Nummer: 76116-20-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR