6-Bromoxindol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

H56012.03 H56012.06
H56012.03EA 62.2 EUR
H56012.03 H56012.06
6-Bromoxindol ≥97%
6-Bromoxindol
Formel: C₈H₆BrNO
Molecular Weight: 212.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179605
CAS-Nummer: 99365-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR