1-Acetyl-5-bromindolin

Lieferant: Thermo Scientific

Synonyme: 1-(5-Bromoindolin-1-yl)ethanone

KM08258DA KM08258EA KM08258.50
MAYBKM08258DAEA 0 EUR
MAYBKM08258DA MAYBKM08258EA MAYBKM08258.50
1-Acetyl-5-bromindolin
1-Acetyl-5-bromindolin
Formel: C₁₀H₁₀BrNO
Molecular Weight: 240,1 g/mol
MDL: MFCD00056017
CAS-Nummer: 22190-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR