Antimon(III)tellurid ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

45922.18 45922.09
45922.18EA 814 EUR
45922.18 45922.09
Antimon(III)tellurid ≥99,999% (Metall-Basis)
Antimon(III)tellurid

Purity: 99.999% CAS No.: 1327-50-0 Molecular Formula: Sb2Te3 Formula Weight: 626.30 Physical Form: -100 Mesh Powder Density: 6.5 Appearance: Grey Odour: Odourless Melting Point: 629° UN No.: 1549 MDL No.: MFCD00016320

Formel: Sb₂Te₃
Molecular Weight: 626.32 g/mol
Schmelzpunkt: 629 °C
Dichte: 6.5
Lagertemperatur: Raumtemperatur
MDL: MFCD00016320
CAS-Nummer: 1327-50-0
EINECS: 215-480-9
UN: 1549
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR