Azetidin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H56351.03
H56351.06
H56351.03EA
140
EUR
H56351.03
H56351.06
Azetidin ≥98%
Azetidin
Formel:
C₃H₇N Molecular Weight: 57,1 g/mol Siedepunkt: 61…62 °C Dichte: 0,847 Flammpunkt: –21 °C (–6 °F) |
MDL:
MFCD00005165 CAS-Nummer: 503-29-7 EINECS: 207-963-8 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...