S-Phenylthioacetat ≥98%
Lieferant: Thermo Scientific
Achtung
|
H55175.06
H55175.14
H55175.06EA
51.9
EUR
H55175.06
H55175.14
S-Phenylthioacetat ≥98%
S-Phenylthioacetat
Purity: 98%
CAS No.: 934-87-2
Molecular Formula: C8H8OS
Formula Weight: 152.21
Flash Point: 79°(174°F)
Physical Form: Liquid
Density: 1.124
Refractive Index: 1.5700
Appearance: Colourless
Boiling Point: 99-100°/6mm
MDL No.: MFCD00008752
Formel:
C₈H₈OS Molecular Weight: 152,22 g/mol Siedepunkt: 99…100 °C (6 mmHg) Dichte: 1,124 Flammpunkt: 79 °C (174 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008752 CAS-Nummer: 934-87-2 EINECS: 213-294-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...